missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Recombinant Human IL-8/CXCL8 (Endothelial Cell Derived)
Beskrivelse
The Recombinant Human IL-8/CXCL8 (Endothelial Cell Derived) is derived from E. coli. The Recombinant Human IL-8/CXCL8 (Endothelial Cell Derived) has been validated for the following applications: Bioactivity.
Tekniske data
Tekniske data
| Formulering | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gen-id (Entrez) | 3576 |
| Molekylvægt (g/mol) | 8.9 kDa |
| Navn | Endothelial Cell-Derived CXCL8/IL-8 aa 23-99 |
| Mængde | 50 μg |
| Kilde | E. coli-derived human IL-8/CXCL8 protein Ala23-Ser99 & Ser28-Ser99 |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1 |
| Konjugeret | Unconjugated |
| Rekombinant | Recombinant |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?