Få mere at vide
R&D Systems™ Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein
Artikelnummer. 18156950
Formulering:
Lyophilized from a 0.2 μm filtered solution in Tris, NaCl, TCEP, EDTA and Tween™ 20 with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in Tris, NaCl, TCEP, EDTA and Tween™ 20 with Trehalose.
Artikelnummer. 18156950
Brand: R&D Systems™ 6995IL010
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
The Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein is derived from E. coli. The Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY
Tekniske data
Formulering Lyophilized from a 0.2 μm filtered solution in Tris, NaCl, TCEP, EDTA and Tween™ 20 with BSA as a carrier protein.
Gen-id (Entrez) 27179
Molekylvægt (g/mol) 17 kDa
Navn IL-36 alpha/IL-1F6 (aa 6-158)
Mængde 10 μg
Kilde E. coli-derived human IL-36 alpha/IL-1F6 protein Lys6-Phe158
Opbevaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Strukturel form Monomer
Konjugeret Unconjugated
Rekombinant Recombinant
Rekonstitution Reconstitute at 100μg/mL in Water.
Renhed eller kvalitetsklasse 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.