Få mere at vide
R&D Systems™ Recombinant Human IL-22 (HEK293-expressed) Protein
Artikelnummer. 18755766
Mængde:
1 mg
10 μg
250 μg
50 μg
Artikelnummer. 18755766
Brand: R&D Systems™ 11311IL050
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Measured by its ability to induce IL-10 secretion in COLO 205 human colorectal adenocarcinoma cells. Nagalakshmi, M.L. et al. (2004) International Immunopharmacology 4:679. The ED50 for this effect is 0.160-1.60 ng/mL.
TRUSTED_SUSTAINABILITY
Tekniske data
Adgangsnummer Q9GZX6.1
Til brug med (applikation) Bioactivity
Formulering Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gen-id (Entrez) 50616
Molekylvægt (g/mol) M.W. Observed: 28-32 kDa, under reducing conditions., M.W. Theoretical: 17 kDa
Mængde 50 μg
Kilde Human embryonic kidney cell, HEK293-derived human IL-22 protein Ala34-Ile179
Opbevaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoksin koncentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
Konjugeret Unconjugated
Rekonstitution Reconstitute the 10 μg size at 100 μg/mL in PBS. Reconstitute all other sizes at 500 μg/mL in PBS.
Renhed eller kvalitetsklasse >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-22