missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ Recombinant Human IL-17E (Human Cell-expressed) Protein

Artikelnummer. 18759086
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18759086 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18759086 Leverandør R&D Systems™ Leverandørnr. 8134IL025

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human IL-17E (Human Cell-expressed) Protein is derived from HEK293. The Recombinant Human IL-17E (Human Cell-expressed) Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Tekniske data

Formulering Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Gen-id (Entrez) 64806
Molekylvægt (g/mol) 17 kDa (monomer)
Navn IL-17E/IL-25
Mængde 25 μg
Kilde Human embryonic kidney cell,HEK293-derived human IL-17E/IL-25 protein Tyr33-Gly177
Opbevaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Strukturel form Disulfide-linked homodimer
Konjugeret Unconjugated
Rekombinant Recombinant
Renhed eller kvalitetsklasse 95%, by SDS-PAGE with silver staining.
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.