missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Recombinant Human IL-17A (Human Cell-expressed) Protein
Beskrivelse
The Recombinant Human IL-17A (Human Cell-expressed) Protein is derived from HEK293. The Recombinant Human IL-17A (Human Cell-expressed) Protein has been validated for the following applications: Bioactivity.
Tekniske data
Tekniske data
| Formulering | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gen-id (Entrez) | 3605 |
| Molekylvægt (g/mol) | 15.1 kDa (monomer) |
| Navn | IL-17/IL-17A |
| Mængde | 25μg |
| Kilde | Human embryonic kidney cell,HEK293-derived human IL-17/IL-17A protein Gly24-Ala155 |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Strukturel form | Disulfide-linked homodimer |
| Gene Alias | CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A |
| Konjugeret | Unconjugated |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?