missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Recombinant Human IL-10 R beta Fc Chimera Protein
Klik for at se tilgængelige muligheder
:
100ug; Carrier Free
Beskrivelse
Measured by its binding ability in a functional ELISA. When Recombinant Human IL 28 R alpha/IFN lambda R1 Fc Chimera (Catalog # 5260-MR ) is immobilized at 5.00 μg/mL (100 μL/well), the concentration of Recombinant Human IL-10 R beta Fc Chimera that produces 50% of the optimal binding response is found to be approximately 0.300-3.00 μg/mL.
Tekniske data
Tekniske data
| Formulering | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gen-id (Entrez) | 3588 |
| Molekylvægt (g/mol) | 51 kDa (monomer) |
| Navn | IL-10 R beta Fc Chimera |
| Mængde | 100μg |
| Kilde | Mouse myeloma cell line,NS0-derived human IL-10 R beta protein, (N-terminus) Human IL-10 R beta (Met20-Ser220) Accession &Num; Q08334 DIEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Strukturel form | Disulfide-linked homodimer |
| Konjugeret | Unconjugated |
| Rekombinant | Recombinant |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion