missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Recombinant Human IL-10 R beta Fc Chimera Avi Protein, CF
Beskrivelse
Measured by its binding ability in a functional ELISA. Biotinylated Recombinant Human IL-10 R beta Fc Chimera Avi-tag (Catalog # AVI11460) binds Recombinant Human IL-28 R alpha /IFN-lambda R1 Fc Chimera (Catalog # 5260-MR ) in the presence of Recombinant Human IL-28B/IFN-lambda 3 (Catalog # 5259-IL ) with an ED50 of 50.0-750 ng/mL.
Tekniske data
Tekniske data
| Til brug med (applikation) | Bioactivity |
| Formulering | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gen-id (Entrez) | 3588 |
| Molekylvægt (g/mol) | M.W.-observed: 78-88 kDa, under reducing conditions, M.W.-theroretical: 52 kDa |
| Mængde | 50 μg |
| Kilde | Chinese Hamster Ovary cell line, CHO-derived human IL-10 R beta protein Human IL-10RB (Met20-Ser220) (N-terminus) Accession # Q08334.2 GGIEGRMD Human IgG1 (Pro100-Lys330) Avi-tag (C-terminus) |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoksin koncentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CDw210b, CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL10R beta, IL-10R2, IL-10R2IL-10R subunit beta, IL10RB, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor subunit 2, interleukin-10 receptor subunit beta, member 4 |
| Konjugeret | Biotin |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?