missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFNA14 / Interferon alpha-14 Protein
A DNA sequence encoding the Homo sapiens (Human) IFNA14 / Interferon alpha-14, was expressed in the hosts and tags indicated.
1185.00 DKK - 17250.00 DKK
Specifications
Accession Number | NP_002163.2 |
---|---|
Gene ID (Entrez) | 3448 |
Name | IFNA14 / Interferon alpha-14 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16061510
|
enQuireBio™
QP6200-ec-10ug |
10 ÎĽg | N/A | ||||||
16001520
|
enQuireBio™
QP6200-ec-50ug |
50 ÎĽg | N/A | ||||||
16051510
|
enQuireBio™
QP6200-ec-100ug |
100 ÎĽg |
3825.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16081510
|
enQuireBio™
QP6200-ec-200ug |
200 ÎĽg | N/A | ||||||
16091510
|
enQuireBio™
QP6200-ec-500ug |
500 ÎĽg | N/A | ||||||
16071510
|
enQuireBio™
QP6200-ec-1mg |
1 mg |
17250.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_002163.2 | |
IFNA14 / Interferon alpha-14 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3448 | |
Research Use Only | |
IFNA14 | |
Recombinant Protein | |
E. coli | |
CNLSQTHSLN NRRTLMLMAQ MRRISPFSCL KDRHDFEFPQ EEFDGNQFQK AQAISVLHEM MQQTFNLFST KNSSAAWDET LLEKFYIELF QQMNDLEACV IQEVGVEETP LMNEDSILAV KKYFQRITLY LMEKKYSPCA WEVVRAEIMR SLSFSTNLQK RLRRKD | |
Greater than 80% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human IFNA14 / Interferon alpha-14 Protein