missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Recombinant Human Hydroxyacid Oxidase-1/HAO-1 Protein
Klik for at se tilgængelige muligheder
Mængde:
10 μg
Beskrivelse
The Recombinant Human Hydroxyacid Oxidase-1/HAO-1 Protein is derived from E. coli. The Recombinant Human Hydroxyacid Oxidase-1/HAO-1 Protein has been validated for the following applications: Enzyme Activity.
Tekniske data
Tekniske data
| Formulering | Supplied as a 0.2μm filtered solution in Sodium Acetate, NaCl and Glycerol. |
| Gen-id (Entrez) | 54363 |
| Molekylvægt (g/mol) | 42 kDa |
| Navn | Hydroxyacid Oxidase-1/HAO-1 |
| Mængde | 10 μg |
| Kilde | E. coli-derived human Hydroxyacid Oxidase-1/HAO-1 protein Leu2-Ile370,with an N-terminal Met and 6-His tag |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Konjugeret | Unconjugated |
| Rekombinant | Recombinant |
| Renhed eller kvalitetsklasse | 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion