missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Hyaluronidase-2 Protein

A DNA sequence encoding the Homo sapiens (Human) Hyaluronidase-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9753-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16068661

  • 4195.00 DKK / 100µg
Estimated Shipment: 03-08-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003764.3
Hyaluronidase-2 Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
8692
Greater than 90% as determined by SDS-PAGE.
Research Use Only
HYAL2
Recombinant Protein
Yeast
MELKPTAPPI FTGRPFVVAW DVPTQDCGPR LKVPLDLNAF DVQASPNEGF VNQNITIFYR DRLGLYPRFD SAGRSVHGGV PQNVSLWAHR KMLQKRVEHY IRTQESAGLA VIDWEDWRPV WVRNWQDKDV YRRLSRQLVA SRHPDWPPDR IVKQAQYEFE FAAQQFMLET LRYVKAVRPR HLWGFYLFPD CYNHDYVQNW ESYTGRCPDV EVARNDQLAW LWAESTALFP SVYLDETLAS SRHGRNFVSF RVQEALRVAR THHANHALPV YVFTRPTYSR RLTGLSEMDL ISTIGESAAL GAAGVILWGD AGYTTSTETC QYLKDYLTRL LVPYVVNVSW ATQYCSRAQC HGHGRCVRRN PSASTFLHLS TNSFRLVPGH APGEPQLRPV GELSWADIDH LQTHFRCQCY LGWSGEQCQW DHRQAAGG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers