missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Hyaluronidase-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Hyaluronidase-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9753-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003764.3 | |
Hyaluronidase-2 Protein | |
Research Use Only | |
HYAL2 | |
Recombinant Protein | |
Yeast | |
MELKPTAPPI FTGRPFVVAW DVPTQDCGPR LKVPLDLNAF DVQASPNEGF VNQNITIFYR DRLGLYPRFD SAGRSVHGGV PQNVSLWAHR KMLQKRVEHY IRTQESAGLA VIDWEDWRPV WVRNWQDKDV YRRLSRQLVA SRHPDWPPDR IVKQAQYEFE FAAQQFMLET LRYVKAVRPR HLWGFYLFPD CYNHDYVQNW ESYTGRCPDV EVARNDQLAW LWAESTALFP SVYLDETLAS SRHGRNFVSF RVQEALRVAR THHANHALPV YVFTRPTYSR RLTGLSEMDL ISTIGESAAL GAAGVILWGD AGYTTSTETC QYLKDYLTRL LVPYVVNVSW ATQYCSRAQC HGHGRCVRRN PSASTFLHLS TNSFRLVPGH APGEPQLRPV GELSWADIDH LQTHFRCQCY LGWSGEQCQW DHRQAAGG | |
Greater than 90% as determined by SDS-PAGE. |
8692 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |