missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSD11B2 Protein
A DNA sequence encoding the Homo sapiens (Human) HSD11B2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9185-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000187.3 | |
HSD11B2 Protein | |
Research Use Only | |
HSD11B2 | |
Recombinant Protein | |
Yeast | |
MERWPWPSGG AWLLVAARAL LQLLRSDLRL GRPLLAALAL LAALDWLCQR LLPPPAALAV LAAAGWIALS RLARPQRLPV ATRAVLITGC DSGFGKETAK KLDSMGFTVL ATVLELNSPG AIELRTCCSP RLRLLQMDLT KPGDISRVLE FTKAHTTSTG LWGLVNNAGH NEVVADAELS PVATFRSCME VNFFGALELT KGLLPLLRSS RGRIVTVGSP AGDMPYPCLG AYGTSKAAVA LLMDTFSCEL LPWGVKVSII QPGCFKTESV RNVGQWEKRK QLLLANLPQE LLQAYGKDYI EHLHGQFLHS LRLAMSDLTP VVDAITDALL AARPRRRYYP GQGLGLMYFI HYYLPEGLRR RFLQAFFISH CLPRALQPGQ PGTTPPQDAA QDPNLSPGPS PAVAR | |
Greater than 90% as determined by SDS-PAGE. |
3291 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |