missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HSD11B2 Protein

A DNA sequence encoding the Homo sapiens (Human) HSD11B2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9185-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16057801

  • 4190.00 DKK / 100µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000187.3
HSD11B2 Protein
Research Use Only
HSD11B2
Recombinant Protein
Yeast
MERWPWPSGG AWLLVAARAL LQLLRSDLRL GRPLLAALAL LAALDWLCQR LLPPPAALAV LAAAGWIALS RLARPQRLPV ATRAVLITGC DSGFGKETAK KLDSMGFTVL ATVLELNSPG AIELRTCCSP RLRLLQMDLT KPGDISRVLE FTKAHTTSTG LWGLVNNAGH NEVVADAELS PVATFRSCME VNFFGALELT KGLLPLLRSS RGRIVTVGSP AGDMPYPCLG AYGTSKAAVA LLMDTFSCEL LPWGVKVSII QPGCFKTESV RNVGQWEKRK QLLLANLPQE LLQAYGKDYI EHLHGQFLHS LRLAMSDLTP VVDAITDALL AARPRRRYYP GQGLGLMYFI HYYLPEGLRR RFLQAFFISH CLPRALQPGQ PGTTPPQDAA QDPNLSPGPS PAVAR
Greater than 90% as determined by SDS-PAGE.
3291
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers