missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSD11B1 Protein
A DNA sequence encoding the Homo sapiens (Human) HSD11B1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6183-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001193670.1 | |
HSD11B1 Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3290 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
HSD11B1 | |
Recombinant Protein | |
E. coli | |
EEFRPEMLQG KKVIVTGASK GIGREMAYHL AKMGAHVVVT ARSKETLQKV VSHCLELGAA SAHYIAGTME DMTFAEQFVA QAGKLMGGLD MLILNHITNT SLNLFHDDIH HVRKSMEVNF LSYVVLTVAA LPMLKQSNGS IVVVSSLAGK VAYPMVAAYS ASKFALDGFF SSIRKEYSVS RVNVSITLCV LGLIDTETAM KAVSGIVHMQ AAPKEECALE IIKGGALRQE EVYYDSSLWT TLLIRNPCRK ILEFLYSTSY NMDRFINK |