missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HMGN4 Protein
A DNA sequence encoding the Homo sapiens (Human) HMGN4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6171-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_006344.1 | |
HMGN4 Protein | |
Research Use Only | |
HMGN4 | |
Recombinant Protein | |
E. coli | |
MPKRKAKGDA KGDKAKVKDE PQRRSARLSA KPAPPKPEPR PKKASAKKGE KLPKGRKGKA DAGKDGNNPA KNRDASTLQS QKAEGTGDAK | |
Greater than 80% as determined by SDS-PAGE. |
10473 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |