missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone chaperone ASF1A Protein
A DNA sequence encoding the Homo sapiens (Human) Histone chaperone ASF1A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8277-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_054753.1 | |
Histone chaperone ASF1A Protein | |
Research Use Only | |
ASF1A | |
Recombinant Protein | |
E. coli | |
MAKVQVNNVV VLDNPSPFYN PFQFEITFEC IEDLSEDLEW KIIYVGSAES EEYDQVLDSV LVGPVPAGRH MFVFQADAPN PGLIPDADAV GVTVVLITCT YRGQEFIRVG YYVNNEYTET ELRENPPVKP DFSKLQRNIL ASNPRVTRFH INWEDNTEKL EDAESSNPNL QSLLSTDALP SASKGWSTSE NSLNVMLESH MDCM | |
Greater than 80% as determined by SDS-PAGE. |
25842 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |