missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone-binding protein RBBP7 Protein
A DNA sequence encoding the Homo sapiens (Human) Histone-binding protein RBBP7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7585-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001185648.1 | |
Histone-binding protein RBBP7 Protein | |
Research Use Only | |
RBBP7 | |
Recombinant Protein | |
E. coli | |
MASKEMFEDT VEERVINEEY KIWKKNTPFL YDLVMTHALQ WPSLTVQWLP EVTKPEGKDY ALHWLVLGTH TSDEQNHLVV ARVHIPNDDA QFDASHCDSD KGEFGGFGSV TGKIECEIKI NHEGEVNRAR YMPQNPHIIA TKTPSSDVLV FDYTKHPAKP DPSGECNPDL RLRGHQKEGY GLSWNSNLSG HLLSASDDHT VCLWDINAGP KEGKIVDAKA IFTGHSAVVE DVAWHLLHES LFGSVADDQK LMIWDTRSNT TSKPSHLVDA HTAEVNCLSF NPYSEFILAT GSADKTVALW DLRNLKLKLH TFESHKDEIF QVHWSPHNET ILASSGTDRR LNVWDLSKIG EEQSAEDAED GPPELLFIHG GHTAKISDFS WNPNEPWVIC SVSEDNIMQI WQMAENIYND EESDVTTSEL EGQGS | |
Greater than 90% as determined by SDS-PAGE. |
5931 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |