missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone-binding protein RBBP7 Protein
A DNA sequence encoding the Homo sapiens (Human) Histone-binding protein RBBP7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7585-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_001185648.1 | |
Histone-binding protein RBBP7 Protein | |
Research Use Only | |
RBBP7 | |
Recombinant Protein | |
E. coli | |
MASKEMFEDT VEERVINEEY KIWKKNTPFL YDLVMTHALQ WPSLTVQWLP EVTKPEGKDY ALHWLVLGTH TSDEQNHLVV ARVHIPNDDA QFDASHCDSD KGEFGGFGSV TGKIECEIKI NHEGEVNRAR YMPQNPHIIA TKTPSSDVLV FDYTKHPAKP DPSGECNPDL RLRGHQKEGY GLSWNSNLSG HLLSASDDHT VCLWDINAGP KEGKIVDAKA IFTGHSAVVE DVAWHLLHES LFGSVADDQK LMIWDTRSNT TSKPSHLVDA HTAEVNCLSF NPYSEFILAT GSADKTVALW DLRNLKLKLH TFESHKDEIF QVHWSPHNET ILASSGTDRR LNVWDLSKIG EEQSAEDAED GPPELLFIHG GHTAKISDFS WNPNEPWVIC SVSEDNIMQI WQMAENIYND EESDVTTSEL EGQGS | |
Greater than 90% as determined by SDS-PAGE. |
5931 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Histone-binding protein RBBP7 Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction