missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HAGH / GLO2 / Glyoxalase II Protein
A DNA sequence encoding the Homo sapiens (Human) HAGH / GLO2 / Glyoxalase II, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8709-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_001035517.1 | |
HAGH / GLO2 / Glyoxalase II Protein | |
Research Use Only | |
HAGH | |
Recombinant Protein | |
E. coli | |
KVEVLPALTD NYMYLVIDDE TKEAAIVDPV QPQKVVDAAR KHGVKLTTVL TTHHHWDHAG GNEKLVKLES GLKVYGGDDR IGALTHKITH LSTLQVGSLN VKCLATPCHT SGHICYFVSK PGGSEPPAVF TGDTLFVAGC GKFYEGTADE MCKALLEVLG RLPPDTRVYC GHEYTINNLK FARHVEPGNA AIREKLAWAK EKYSIGEPTV PSTLAEEFTY NPFMRVREKT VQQHAGETDP VTTMRAVRRE KDQFKMPRD | |
Greater than 90% as determined by SDS-PAGE. |
3029 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human HAGH / GLO2 / Glyoxalase II Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction