missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GTF2E2 Protein
A DNA sequence encoding the Homo sapiens (Human) GTF2E2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6131-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002086.1 | |
GTF2E2 Protein | |
Research Use Only | |
GTF2E2 | |
Recombinant Protein | |
E. coli | |
MDPSLLRERE LFKKRALSTP VVEKRSASSE SSSSSSKKKK TKVEHGGSSG SKQNSDHSNG SFNLKALSGS SGYKFGVLAK IVNYMKTRHQ RGDTHPLTLD EILDETQHLD IGLKQKQWLM TEALVNNPKI EVIDGKYAFK PKYNVRDKKA LLRLLDQHDQ RGLGGILLED IEEALPNSQK AVKALGDQIL FVNRPDKKKI LFFNDKSCQF SVDEEFQKLW RSVTVDSMDE EKIEEYLKRQ GISSMQESGP KKVAPIQRRK KPASQKKRRF KTHNEHLAGV LKDYSDITSS K | |
Greater than 90% as determined by SDS-PAGE. |
2961 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |