Learn More
R&D Systems™ Recombinant Human Growth Hormone (GH) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 1067-GH-025
This item is not returnable.
View return policy
Description
The Recombinant Human Growth Hormone (GH) Protein is derived from E. coli. The Recombinant Human Growth Hormone (GH) Protein has been validated for the following applications: Bioactivity.
Specifications
Lyophilized from a 0.2ÎĽm filtered solution in Tris and NaCl with BSA as a carrier protein. | |
22 kDa | |
25 ÎĽg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
2688 | |
Growth Hormone | |
E. coli-derived human Growth Hormone protein Phe27-Phe217,with an N-terminal Met | |
GH1, GHB5, GHIGHD1B, GHN, GH-N, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N, IGHD1A, IGHD2, Somatotropin | |
Recombinant |
Product Content Correction
Product Title
R&D Systems™ Recombinant Human Growth Hormone (GH) Protein > 25ug
Spot an opportunity for improvement?Share a Content Correction