missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GNB5 Protein
A DNA sequence encoding the Homo sapiens (Human) GNB5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6103-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006569.1 | |
GNB5 Protein | |
Research Use Only | |
GNB5 | |
Recombinant Protein | |
E. coli | |
MATEGLHENE TLASLKSEAE SLKGKLEEER AKLHDVELHQ VAERVEALGQ FVMKTRRTLK GHGNKVLCMD WCKDKRRIVS SSQDGKVIVW DSFTTNKEHA VTMPCTWVMA CAYAPSGCAI ACGGLDNKCS VYPLTFDKNE NMAAKKKSVA MHTNYLSACS FTNSDMQILT ASGDGTCALW DVESGQLLQS FHGHGADVLC LDLAPSETGN TFVSGGCDKK AMVWDMRSGQ CVQAFETHES DINSVRYYPS GDAFASGSDD ATCRLYDLRA DREVAIYSKE SIIFGASSVD FSLSGRLLFA GYNDYTINVW DVLKGSRVSI LFGHENRVST LRVSPDGTAF CSGSWDHTLR VWA | |
Greater than 90% as determined by SDS-PAGE. |
10681 | |
200 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |