Learn More
enQuireBio™ Recombinant Human Glutathione S-transferase A4 Protein
A DNA sequence encoding the Homo sapiens (Human) Glutathione S-transferase A4, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6124-ec-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16070980
  • 3450.00 DKK / 100µg
Estimated Shipment: 08-07-2024
to see stock.
Specifications
NP_001503.1
Glutathione S-transferase A4 Protein
Research Use Only
GSTA4
Recombinant Protein
E. coli
MAARPKLHYP NGRGRMESVR WVLAAAGVEF DEEFLETKEQ LYKLQDGNHL LFQQVPMVEI DGMKLVQTRS ILHYIADKHN LFGKNLKERT LIDMYVEGTL DLLELLIMHP FLKPDDQQKE VVNMAQKAII RYFPVFEKIL RGHGQSFLVG NQLSLADVIL LQTILALEEK IPNILSAFPF LQEYTVKLSN IPTIKRFLEP GSKKKPPPDE IYVRTVYNIF RP
Greater than 90% as determined by SDS-PAGE.
2941
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol