missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glutamate receptor 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Glutamate receptor 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6120-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000819.3 | |
Glutamate receptor 3 Protein | |
Research Use Only | |
GRIA3 | |
Recombinant Protein | |
Yeast | |
SLLGHYKWEK FVYLYDTERG FSILQAIMEA AVQNNWQVTA RSVGNIKDVQ EFRRIIEEMD RRQEKRYLID CEVERINTIL EQVVILGKHS RGYHYMLANL | |
Greater than 90% as determined by SDS-PAGE. |
2892 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |