missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GH1 / Growth hormone 1 Protein
A DNA sequence encoding the Human GH1 / Growth hormone 1, was expressed in the hosts and tags indicated.
780.00 DKK - 2300.00 DKK
Specifications
Accession Number | NP_000506.2 |
---|---|
Gene ID (Entrez) | 2688 |
Name | GH1 / Growth hormone 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than 1.0 EU/ug as determined by LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15902908
|
enQuireBio™
QP10147-EC-20UG |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15992898
|
enQuireBio™
QP10147-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000506.2 | |
GH1 / Growth hormone 1 Protein | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
FPTIPLSRLF DNAMLRAHRL HQLAFDTYQE FEEAYIPKEQ KYSFLQNPQT SLCFSESIPT PSNREETQQK SNLELLRISL LLIQSWLEPV QFLRSVFANS LVYGASDSNV YDLLKDLEEG IQTLMGRLED GSPRTGQIFK QTYSKFDTNS HNDDALLKNY GLLYCFRKDM DKVETFLRIV QCRSVEGSCG F | |
Greater than 98% as determined by SDS-PAGE and HPLC. |
2688 | |
Research Use Only | |
GH1 | |
The ED50 as determined by a cell proliferation assay using rat Nb2-11 lymphoma cells is less than 0.1 ng/ml, corresponding to a specific activity of > 1.0 M 107 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |