missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ Recombinant Human GDF-15 (Human Cell-expressed) Protein

Artikelnummer. 18739177
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 μg
25 μg
Formulering:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Formulering
18739177 25 μg Lyophilized from a 0.2μm filtered solution in HCl.
18188150 10 μg Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
18146600 10 μg Lyophilized from a 0.2μm filtered solution in HCl.
18727957 25 μg Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18739177 Leverandør R&D Systems™ Leverandørnr. 8146GD025/CF

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Measured by its ability to activate SRE-SEAP reporter in HEK293 human embryonic kidney cells transfected with human c-Ret and human GFRAL. The ED50 for this effect is 0.2-2 ng/mL.

The Recombinant Human GDF-15 (Human Cell-expressed) Protein is derived from HEK293.
TRUSTED_SUSTAINABILITY

Tekniske data

Formulering Lyophilized from a 0.2μm filtered solution in HCl.
Gen-id (Entrez) 9518
Molekylvægt (g/mol) 12 kDa (monomer)
Navn GDF-15
Mængde 25 μg
Kilde Human embryonic kidney cell,HEK293-derived human GDF-15 protein Ala197-Ile308
Opbevaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Strukturel form Disulfide-linked homodimer
Konjugeret Unconjugated
Rekombinant Recombinant
Renhed eller kvalitetsklasse 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.