missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Recombinant Human Galectin-3 (Human Cell-expressed)
Beskrivelse
Measured by its ability to support the adhesion of MOLT-4 human acute lymphoblastic leukemia cells. The ED50 for this effect id 0.6-3 μg/mL. Measured by its ability to agglutinate human red blood cells. Hadari, Y.R. et al. (2000) J. Cell Sci. 113:2385. The ED50 for this effect is 3-10 μg/mL.
Ordreinfo
The product is shipped at ambient temperature. Upon receipt, store it immediately
Tekniske data
Tekniske data
| Adgangsnummer | P17931 |
| Til brug med (applikation) | Bioactivity |
| Formulering | Lyophilized from a 0.2 μm filtered solution in HEPES, NaCl, TCEP, PEG and Trehalose. |
| Gen-id (Entrez) | 3958 |
| Molekylvægt (g/mol) | M.W. (Observed): 26-31 kDa, reducing conditions; M.W. (theoretical): 25 kDa |
| Navn | Galectin-3, Carrier Free |
| Mængde | 50 μg |
| Kilde | Human embryonic kidney cell, HEK293-derived human Galectin-3 protein Ala2-Ile250 |
| Immunogen | Ala2-Ile250 |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Vis mere |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?