missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Galactocerebrosidase Protein
A DNA sequence encoding the Homo sapiens (Human) Galactocerebrosidase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9145-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000144.2 | |
Galactocerebrosidase Protein | |
Research Use Only | |
GALC | |
Recombinant Protein | |
Yeast | |
YVLDDSDGLG REFDGIGAVS GGGATSRLLV NYPEPYRSQI LDYLFKPNFG ASLHILKVEI GGDGQTTDGT EPSHMHYALD ENYFRGYEWW LMKEAKKRNP NITLIGLPWS FPGWLGKGFD WPYVNLQLTA YYVVTWIVGA KRYHDLDIDY IGIWNERSYN ANYIKILRKM LNYQGLQRVK IIASDNLWES ISASMLLDAE LFKVVDVIGA HYPGTHSAKD AKLTGKKLWS SEDFSTLNSD MGAGCWGRIL NQNYINGYMT STIAWNLVAS YYEQLPYGRC GLMTAQEPWS GHYVVESPVW VSAHTTQFTQ PGWYYLKTVG HLEKGGSYVA LTDGLGNLTI IIETMSHKHS KCIRPFLPYF NVSQQFATFV LKGSFSEIPE LQVWYTKLGK TSERFLFKQL DSLWLLDSDG SFTLSLHEDE LFTLTTLTTG RKGSYPLPPK SQPFPSTYKD DFNVDYPFFS EAPNFADQTG VFEYFTNIED PGEHHFTLRQ VLNQRPITWA ADASNTISII GDYNWTNLTI KCDVYIETPD TGGVFIAGRV NKGGILIRSA RGIFFWIFAN GSYRVTGDLA GWIIYALGRV EVTAKKWYTL TLTIKGHFAS GMLNDKSLWT DIPVNFPKNG WAAIGTHSFE FAQFDNFLVE ATR | |
Greater than 90% as determined by SDS-PAGE. |
2581 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |