missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Galactocerebrosidase Protein

A DNA sequence encoding the Homo sapiens (Human) Galactocerebrosidase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9145-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16067701

  • 6005.00 DKK / 100µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000144.2
Galactocerebrosidase Protein
Research Use Only
GALC
Recombinant Protein
Yeast
YVLDDSDGLG REFDGIGAVS GGGATSRLLV NYPEPYRSQI LDYLFKPNFG ASLHILKVEI GGDGQTTDGT EPSHMHYALD ENYFRGYEWW LMKEAKKRNP NITLIGLPWS FPGWLGKGFD WPYVNLQLTA YYVVTWIVGA KRYHDLDIDY IGIWNERSYN ANYIKILRKM LNYQGLQRVK IIASDNLWES ISASMLLDAE LFKVVDVIGA HYPGTHSAKD AKLTGKKLWS SEDFSTLNSD MGAGCWGRIL NQNYINGYMT STIAWNLVAS YYEQLPYGRC GLMTAQEPWS GHYVVESPVW VSAHTTQFTQ PGWYYLKTVG HLEKGGSYVA LTDGLGNLTI IIETMSHKHS KCIRPFLPYF NVSQQFATFV LKGSFSEIPE LQVWYTKLGK TSERFLFKQL DSLWLLDSDG SFTLSLHEDE LFTLTTLTTG RKGSYPLPPK SQPFPSTYKD DFNVDYPFFS EAPNFADQTG VFEYFTNIED PGEHHFTLRQ VLNQRPITWA ADASNTISII GDYNWTNLTI KCDVYIETPD TGGVFIAGRV NKGGILIRSA RGIFFWIFAN GSYRVTGDLA GWIIYALGRV EVTAKKWYTL TLTIKGHFAS GMLNDKSLWT DIPVNFPKNG WAAIGTHSFE FAQFDNFLVE ATR
Greater than 90% as determined by SDS-PAGE.
2581
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers