missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FNDC5 Protein
A DNA sequence encoding the Homo sapiens (Human) FNDC5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7882-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001165411.2 | |
FNDC5 Protein | |
Research Use Only | |
FNDC5 | |
Recombinant Protein | |
E. coli | |
DSPSAPVNVT VRHLKANSAV VSWDVLEDEV VIGFAISQQK KDVRMLRFIQ EVNTTTRSCA LWDLEEDTEY IVHVQAISIQ GQSPASEPVL FKTPREAEKM ASKNKDEVTM KE | |
Greater than 90% as determined by SDS-PAGE. |
252995 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |