Learn More
enQuireBio™ Recombinant Human FGF21 / Fibroblast Growth Factor 21 Protein
Recombinant Protein
Brand:  enQuireBio™ QP5272-500ug
 View more versions of this product
Product Code. 15915819
  • 18805.00 DKK / 500µg
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
NP_061986.1
FGF21 / Fibroblast Growth Factor 21 Protein
Research Use Only
FGF21
Human
Untagged
10 mM sodium phosphate, 100 mM soldium chloride, pH 7.5
26291
500 ÎĽg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
MHPIPDSSPL . LQFGGQVRQ R. YLYTDDAQ QT. EAHLEIR EDG. TVGGAA DQSP. ESLLQ LKALK. PGVI QILGVK. TSR FLCQRPD. GA LYGSLHFD. P EACSFRELLL EDGYNVYQS.  EAHGLPLHLP . GNKSPHRDP A. PRGPARFL PL. PGLPPAL PEP. PGILAP QPPD. VGSSD PLSMV. GPSQ GRSPSY. AS
Greater than 95% as determined by SDS-PAGE.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human FGF21 / Fibroblast Growth Factor 21 Protein > 500ÎĽg