missing translation for 'onlineSavingsMsg'
Få mere at vide

R&DSystems™ Recombinant Human FGF basic Heat Stable Protein, CF

Artikelnummer. 30067256
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
50 μg
500 μg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 30067256

Brand: R&DSystems™ BTFGFBHS100

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Recombinant Protein

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is 0.0500-0.600 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Adgangsnummer NP_001997.5
Til brug med (applikation) Bioactivity
Formulering Lyophilized from a 0.2 μm filtered solution in HEPES and Sodium Sulfate with Trehalose.
Gen-id (Entrez) 2247
Molekylvægt (g/mol) M.W.-observed: 16-19 kDa, under reducing conditions., M.W.-theroretical: 17 kDa
Mængde 100 μg
Kilde E. coli-derived human FGF basic/FGF2/bFGF protein Ala135-Ser288 (with modifications)
Opbevaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoksin koncentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Konjugeret Unconjugated
Rekonstitution Reconstitute 10 μg size at 100 μg/mL in sterile deionized water. Reconstitute all the other sizes at 500 μg/mL in sterile deionized water.
Protein FGF basic/FGF2/bFGF
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.