missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fc receptor-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Fc receptor-like protein 6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7695-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001004310.2 | |
Fc receptor-like protein 1 | |
Research Use Only | |
FCRL6 | |
Recombinant Protein | |
E. coli | |
LYLQAWPNPV FEGDALTLRC QGWKNTPLSQ VKFYRDGKFL HFSKENQTLS MGAATVQSRG QYSCSGQVMY IPQTFTQTSE TAMVQVQELF PPPVLSAIPS PEPREGSLVT LRCQTKLHPL RSALRLLFSF HKDGHTLQDR GPHPELCIPG AKEGDSGLYW CEVAPEGGQV QKQSPQLEVR VQAPVSRPVL TLHHGPADPA VGDMVQLLCE AQRGSPPILY SFYLDEKIVG NHSAPCGGTT SLLFPVKSEQ DAGNYSCEAE NSVSRERSEP KKLSLKGSQV LFTPASNW | |
Greater than 90% as determined by SDS-PAGE. |
343413 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |