Beskrivelse
Recombinant protein for Human FABP4/A-FABP
The Recombinant Human FABP4/A-FABP Protein is derived from E. coli. The Recombinant Human FABP4/A-FABP Protein has been validated for the following applications: SDS-Page.
The Recombinant Human FABP4/A-FABP Protein is derived from E. coli. The Recombinant Human FABP4/A-FABP Protein has been validated for the following applications: SDS-Page.
Tekniske data
Tekniske data
| Koncentration | Lyoph |
| Til brug med (applikation) | SDS-PAGE |
| Gen-id (Entrez) | 2167 |
| Molekylvægt (g/mol) | 16 kDa |
| Mængde | 100 μg |
| Kilde | E. coli |
| Opbevaringskrav | Store at -20 to -80°C. Avoid freeze-thaw cycles. |
| Regulatorisk status | RUO |
| Endotoksin koncentration | <0.1 ng/ug |
| Gen symbol | FABP4 |
| Vis mere |