missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ESAM / Endothelial Cell Adhesion Molecule Protein
A DNA sequence encoding the Homo sapiens (Human) ESAM / Endothelial Cell Adhesion Molecule, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7960-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_620411.2 | |
ESAM / Endothelial Cell Adhesion Molecule Protein | |
Research Use Only | |
ESAM | |
Recombinant Protein | |
E. coli | |
QLQLHLPANR LQAVEGGEVV LPAWYTLHGE VSSSQPWEVP FVMWFFKQKE KEDQVLSYIN GVTTSKPGVS LVYSMPSRNL SLRLEGLQEK DSGPYSCSVN VQDKQGKSRG HSIKTLELNV LVPPAPPSCR LQGVPHVGAN VTLSCQSPRS KPAVQYQWDR QLPSFQTFFA PALDVIRGSL SLTNLSSSMA GVYVCKAHNE VGTAQCNVTL EVSTGPGAA | |
Greater than 90% as determined by SDS-PAGE. |
90952 | |
500 ÎĽg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human ESAM / Endothelial Cell Adhesion Molecule Protein > 500μg
Spot an opportunity for improvement?Share a Content Correction