Få mere at vide
R&D Systems™ Recombinant Human EphB4 Fc Chimera Protein
Artikelnummer. 18767276
Mængde:
100 μg
Artikelnummer. 18767276
Brand: R&D Systems™ 11307B4100
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Measured by its binding ability in a functional ELISA. When Recombinant Human EphB4 Fc Chimera (Catalog # 11307-B4) is immobilized at 0.3 μg/mL (100 μL/well), Recombinant Mouse Ephrin-B2 Fc Chimera (Catalog # 496-EB ) binds with an ED50 of 0.600-7.20 ng/mL.
TRUSTED_SUSTAINABILITY
Tekniske data
Adgangsnummer AAH52804.1
Til brug med (applikation) Bioactivity
Formulering Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gen-id (Entrez) 2050
Molekylvægt (g/mol) M.W. Observed: 95-108 kDa, under reducing conditions., M.W. Theoretical: 58 kDa
Mængde 100 μg
Kilde Mouse myeloma cell line, NS0-derived human EphB4 protein Human EphB4 (Leu16-Arg539) (N-terminus) Accession # AAH52804.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Opbevaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoksin koncentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias EC 2.7.10, EC 2.7.10.1, EPH receptor B4, EphB4, ephrin type-B receptor 4, hepatoma transmembrane kinase, Htk, HTKephrin receptor EphB4, Mdk2, Myk1, soluble EPHB4 variant 1, soluble EPHB4 variant 2, soluble EPHB4 variant 3, Tyro11, Tyrosine-protein kinase receptor HTK, Tyrosine-protein kinase TYRO11
Konjugeret Unconjugated
Rekonstitution Reconstitute at 500 μg/mL in PBS.
Renhed eller kvalitetsklasse >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein EphB4