missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Envelope glycoprotein 1
A DNA sequence encoding the Homo sapiens (Human) Envelope glycoprotein H, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7033-ec-100ug
This item is not returnable.
View return policy
Specifications
Envelope glycoprotein 1 | |
Research Use Only | |
gH | |
Recombinant Protein | |
E. coli | |
RYGADAASEA LDPHAFHLLL NTYGRPIRFL RENTTQCTYN SSLRNSTVVR ENAISFNFFQ SYNQYYVFHM PRCLFAGPLA EQFLNQVDLT ETLERYQQRL NTYALVSKDL ASYRSFSQQL KAQDSLGQQP TTVPPPIDLS IPHVWMPPQT TPHDWKGSHT TSGLHRPHFN QT | |
Greater than 90% as determined by SDS-PAGE. |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Envelope glycoprotein 1 > 100μg
Spot an opportunity for improvement?Share a Content Correction