missing translation for 'onlineSavingsMsg'
Få mere at vide

enQuireBio™ Recombinant Human Elongation factor Tu, mitochondrial Protein

Artikelnummer. 16066320
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 16066320

Brand: enQuireBio™ QP6855ye100ug

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

A DNA sequence encoding the Homo sapiens (Human) Elongation factor Tu, mitochondrial, was expressed in the hosts and tags indicated.

Specifications

Adgangsnummer NP_003312.3
Gen-id (Entrez) 7284
Navn Elongation factor Tu, mitochondrial Protein
Mængde 100 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol TUFM
Protein længde Full Length
Produkttype Recombinant Protein
Krydsreaktivitet Human
Arter Yeast
Protein tag His
Sekvens AVEAKKTYVR DKPHVNVGTI GHVDHGKTTL TAAITKILAE GGGAKFKKYE EIDNAPEERA RGITINAAHV EYSTAARHYA HTDCPGHADY VKNMITGTAP LDGCILVVAA NDGPMPQTRE HLLLARQIGV EHVVVYVNKA DAVQDSEMVE LVELEIRELL TEFGYKGEET PVIVGSALCA LEGRDPELGL KSVQKLLDAV DTYIPVPARD LEKPFLLPVE AVYSVPGRGT VVTGTLERGI LKKGDECELL GHSKNIRTVV TGIEMFHKSL ERAEAGDNLG ALVRGLKRED LRRGLVMVKP GSIKPHQKVE AQVYILSKEE GGRHKPFVSH FMPVMFSLTW DMACRIILPP EKELAMPGED LKFNLILRQP MILEKGQRFT LRDGNRTIGT GLVTNTLAMT EEEKNIKWG
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.