Få mere at vide
enQuireBio™ Recombinant Human Elongation factor Tu, mitochondrial Protein
Artikelnummer. p-7153707
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
Artikelnummer. 16076320
Brand: enQuireBio™ QP6855ye10ug
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
A DNA sequence encoding the Homo sapiens (Human) Elongation factor Tu, mitochondrial, was expressed in the hosts and tags indicated.
Tekniske data
Adgangsnummer NP_003312.3
Gen-id (Entrez) 7284
Navn Elongation factor Tu, mitochondrial Protein
Mængde 10 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol TUFM
Protein længde Full Length
Produkttype Recombinant Protein
Krydsreaktivitet Human
Arter Yeast
Protein tag His
Sekvens AVEAKKTYVR DKPHVNVGTI GHVDHGKTTL TAAITKILAE GGGAKFKKYE EIDNAPEERA RGITINAAHV EYSTAARHYA HTDCPGHADY VKNMITGTAP LDGCILVVAA NDGPMPQTRE HLLLARQIGV EHVVVYVNKA DAVQDSEMVE LVELEIRELL TEFGYKGEET PVIVGSALCA LEGRDPELGL KSVQKLLDAV DTYIPVPARD LEKPFLLPVE AVYSVPGRGT VVTGTLERGI LKKGDECELL GHSKNIRTVV TGIEMFHKSL ERAEAGDNLG ALVRGLKRED LRRGLVMVKP GSIKPHQKVE AQVYILSKEE GGRHKPFVSH FMPVMFSLTW DMACRIILPP EKELAMPGED LKFNLILRQP MILEKGQRFT LRDGNRTIGT GLVTNTLAMT EEEKNIKWG
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 90% as determined by SDS-PAGE.