missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EIF3M Protein

A DNA sequence encoding the Homo sapiens (Human) EIF3M, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7743-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16070531

  • 1365.00 DKK / 10µg
Estimated Shipment: 31-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001294858.1
EIF3M Protein
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
10480
Greater than 90% as determined by SDS-PAGE.
Research Use Only
EIF3M
Recombinant Protein
E. coli
SVPAFIDISE EDQAAELRAY LKSKGAEISE ENSEGGLHVD LAQIIEACDV CLKEDDKDVE SVMNSVVSLL LILEPDKQEA LIESLCEKLV KFREGERPSL RLQLLSNLFH GMDKNTPVRY TVYCSLIKVA ASCGAIQYIP TELDQVRKWI SDWNLTTEKK HTLLRLLYEA LVDCKKSDAA SKVMVELLGS YTEDNASQAR VDAHRCIVRA LKDPNAFLFD HLLTLKPVKF LEGELIHDLL TIFVSAKLAS YVKFYQNNKD FIDSLGLLHE QNMAKMRLLT FMGMAVENKE ISFDTMQQEL QIGADDVEAF VIDAVRTKMV YCKIDQTQRK VVVSHSTHRT FGKQQWQQLY DTLNAWKQNL NKVKNSLLSL SDT
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers