missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase MIB1 Protein

A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase MIB1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8760-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16086331

  • 15565.00 DKK / 1mg
Estimated Shipment: 09-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_065825.1
E3 ubiquitin-protein ligase MIB1 Protein
Research Use Only
MIB1
Recombinant Protein
E. coli
RNNRVMVEGV GARVVRGPDW KWGKQDGGEG HVGTVRSFES PEEVVVVWDN GTAANYRCSG AYDLRILDSA PTGIKHDGTM CDTCRQQPII GIRWKCAECT NYDLCTVCYH GDKHHLRHRF YRITTPGSER VLLESRRKSK KITARGIFAG ARVVRGVDWQ WEDQDGGNGR RGKVTEIQDW SASSPHSAAY VLWDNGAKNL YRVGFEGMSD LKCVQDAKGG SFYRDHCPVL GEQNGNRNPG GLQIGDLVNI DLDLEIVQSL QHGHGGWTDG MFETLTTTGT VCGIDEDHDI VVQYPSGNRW TFNPAVLTKA NIVRSGDAAQ GAEGGTSQ
Greater than 90% as determined by SDS-PAGE.
57534
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers