missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dystonin Protein
A DNA sequence encoding the Homo sapiens (Human) Dystonin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7837-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001138241.1 | |
Dystonin Protein | |
Research Use Only | |
DST | |
Recombinant Protein | |
Yeast | |
MAGYLSPAAY LYVEEQEYLQ AYEDVLERYK DERDKVQKKT FTKWINQHLM KVRKHVNDLY EDLRDGHNLI SLLEVLSGDT LPREKGRMRF HRLQNVQIAL DYLKRRQVKL VNIRNDDITD GNPKLTLGLI WTIILHFQIS DIHVTGESED MSAKERLLLW TQQATEGYAG IRCENFTTCW RDGKLFNAII HKYRP | |
Greater than 90% as determined by SDS-PAGE. |
667 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |