Learn More
enQuireBio™ Recombinant Human DYDC2 / DPY30 domain containing 2 Protein
A DNA sequence encoding the Homo sapiens (Human) DYDC2 / DPY30 domain containing 2, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7875-ec-500ug
 View more versions of this product
Product Code. 16001321
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
NP_001256970.1
DYDC2 / DPY30 domain containing 2 Protein
Research Use Only
DYDC2
Recombinant Protein
E. coli
METNYLKRCF GNCLAQALAE VAKVRPSDPI EYLAHWLYHY RKTAKAKEEN REKKIHLQEE YDSSLKEMEM TEMLKQEEYQ IQQNCEKCHK ELTSETVSTK KTIFMQEDTN PLEKEALKQE FLPGTSSLIP GMPQQVPPSE SAGQIDQNFK MPQEINYKEA FQHEVAHEMP PGSKSPF
Greater than 90% as determined by SDS-PAGE.
84332
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Certificates
Promotions
Promotions
Product Content Correction
Product Title
enQuireBio™ Recombinant Human DYDC2 / DPY30 domain containing 2 Protein > 500μg