missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA repair protein XRCC3 Protein
A DNA sequence encoding the Homo sapiens (Human) DNA repair protein XRCC3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6893-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001093588.1 | |
DNA repair protein XRCC3 Protein | |
Research Use Only | |
XRCC3 | |
Recombinant Protein | |
E. coli | |
MDLDLLDLNP RIIAAIKKAK LKSVKEVLHF SGPDLKRLTN LSSPEVWHLL RTASLHLRGS SILTALQLHQ QKERFPTQHQ RLSLGCPVLD ALLRGGLPLD GITELAGRSS AGKTQLALQL CLAVQFPRQH GGLEAGAVYI CTEDAFPHKR LQQLMAQQPR LRTDVPGELL QKLRFGSQIF IEHVADVDTL LECVNKKVPV LLSRGMARLV VIDSVAAPFR CEFDSQASAP RARHLQSLGA TLRELSSAFQ SPVLCINQVT EAMEEQGAAH GPLGFWDERV SPALGITWAN QLLVRLLADR LREEEAALGC PARTLR | |
Greater than 90% as determined by SDS-PAGE. |
7517 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |