missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DNA repair protein XRCC3 Protein

A DNA sequence encoding the Homo sapiens (Human) DNA repair protein XRCC3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6893-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16066600

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001093588.1
DNA repair protein XRCC3 Protein
Research Use Only
XRCC3
Recombinant Protein
E. coli
MDLDLLDLNP RIIAAIKKAK LKSVKEVLHF SGPDLKRLTN LSSPEVWHLL RTASLHLRGS SILTALQLHQ QKERFPTQHQ RLSLGCPVLD ALLRGGLPLD GITELAGRSS AGKTQLALQL CLAVQFPRQH GGLEAGAVYI CTEDAFPHKR LQQLMAQQPR LRTDVPGELL QKLRFGSQIF IEHVADVDTL LECVNKKVPV LLSRGMARLV VIDSVAAPFR CEFDSQASAP RARHLQSLGA TLRELSSAFQ SPVLCINQVT EAMEEQGAAH GPLGFWDERV SPALGITWAN QLLVRLLADR LREEEAALGC PARTLR
Greater than 90% as determined by SDS-PAGE.
7517
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers