missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Deoxyribonuclease gamma Protein
A DNA sequence encoding the Homo sapiens (Human) Deoxyribonuclease gamma, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7580-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001243489.1 | |
Deoxyribonuclease gamma Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
1776 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
DNASE1L3 | |
Recombinant Protein | |
E. coli | |
MRICSFNVRS FGESKQEDKN AMDVIVKVIK RCDIILVMEI KDSNNRICPI LMEKLNRNSR RGITYNYVIS SRLGRNTYKE QYAFLYKEKL VSVKRSYHYH DYQDGDADVF SREPFVVWFQ SPHTAVKDFV IIPLHTTPET SVKEIDELVE VYTDVKHRWK AENFIFMGDF NAGCSYVPKK AWKNIRLRTD PRFVWLIGDQ EDTTVKKSTN CAYDRIVLRG QEIVSSVVPK SNSVFDFQKA YKLTEEEALD VSDHFPVEFK LQSSRAFTNS KKSVTLRKKT KSKRS |