Learn More
enQuireBio™ Recombinant Human DDB1- and CUL4-associated factor 4 Protein
A DNA sequence encoding the Homo sapiens (Human) DDB1- and CUL4-associated factor 4, was expressed in the hosts and tags indicated.
1185.00 DKK - 17250.00 DKK
Specifications
Accession Number | NP_001156980.1 |
---|---|
Gene ID (Entrez) | 26094 |
Name | DDB1- and CUL4-associated factor 4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16062041
|
enQuireBio™
QP7992-ec-10ug |
10 ÎĽg | N/A | ||||||
16002051
|
enQuireBio™
QP7992-ec-50ug |
50 ÎĽg | N/A | ||||||
16052041
|
enQuireBio™
QP7992-ec-100ug |
100 ÎĽg |
3825.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16082041
|
enQuireBio™
QP7992-ec-200ug |
200 ÎĽg | N/A | ||||||
16092041
|
enQuireBio™
QP7992-ec-500ug |
500 ÎĽg | N/A | ||||||
16072041
|
enQuireBio™
QP7992-ec-1mg |
1 mg |
17250.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001156980.1 | |
DDB1- and CUL4-associated factor 4 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
26094 | |
Research Use Only | |
DCAF4 | |
Recombinant Protein | |
E. coli | |
MNKSRWQSRR RHGRRSHQQN PWFRLRDSED RSDSRAAQPA HDSGHGDDES PSTSSGTAGT SSVPELPGFY FDPEKKRYFR LLPGHNNCNP LTKESIRQKE MESKRLRLLQ EEDRRKKIAR MGFNASSMLR KSQLGFLNVT NYCHLAHELR LSCMERKKVQ IRSMDPSALA SDRFNLILAD TNSDRLFTVN DVKVGGSKYG IINLQSLKTP TLKVFMHENL YFTNRKVNSV CWASLNHLDS HILLCLMGLA ETPGCATLLP ASLFVNSHPG IDRPGMLCSF RIPGAWSCAW SLNIQANNCF STGLSRRVLL TNVVTGHRQS FGTNSDVLAQ QFALMAPLLF NGCRSGEIFA IDLRCGNQGK GWKATRLFHD SAVTSVRILQ DEQYLMASDM AGKIKLWDLR TTKCVRQYEG HVNEYAYLPL HVHEEEGILV AVGQDCYTRI WSLHDARLLR TIPSPYPASK ADIPSVAFSS RLGGSRGAPG LLMAVGQDLY CYSYS | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
enQuireBio™ Recombinant Human DDB1- and CUL4-associated factor 4 Protein