missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cytosolic 5'-nucleotidase 1A Protein

A DNA sequence encoding the Homo sapiens (Human) Cytosolic 5'-nucleotidase 1A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9892-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16088931

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_115915.1
Cytosolic 5'-nucleotidase 1A Protein
Research Use Only
NT5C1A
Recombinant Protein
Yeast
MEPGQPREPQ EPREPGPGAE TAAAPVWEEA KIFYDNLAPK KKPKSPKPQN AVTIAVSSRA LFRMDEEQQI YTEQGVEEYV RYQLEHENEP FSPGPAFPFV KALEAVNRRL RELYPDSEDV FDIVLMTNNH AQVGVRLINS INHYDLFIER FCMTGGNSPI CYLKAYHTNL YLSADAEKVR EAIDEGIAAA TIFSPSRDVV VSQSQLRVAF DGDAVLFSDE SERIVKAHGL DRFFEHEKAH ENKPLAQGPL KGFLEALGRL QKKFYSKGLR LECPIRTYLV TARSAASSGA RALKTLRSWG LETDEALFLA GAPKGPLLEK IRPHIFFDDQ MFHVAGAQEM GTVAAHVPYG VAQTPRRTAP AKQAPSAQ
Greater than 90% as determined by SDS-PAGE.
84618
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers