missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytosolic 5'-nucleotidase 1A Protein
A DNA sequence encoding the Homo sapiens (Human) Cytosolic 5'-nucleotidase 1A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9892-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_115915.1 | |
Cytosolic 5'-nucleotidase 1A Protein | |
Research Use Only | |
NT5C1A | |
Recombinant Protein | |
Yeast | |
MEPGQPREPQ EPREPGPGAE TAAAPVWEEA KIFYDNLAPK KKPKSPKPQN AVTIAVSSRA LFRMDEEQQI YTEQGVEEYV RYQLEHENEP FSPGPAFPFV KALEAVNRRL RELYPDSEDV FDIVLMTNNH AQVGVRLINS INHYDLFIER FCMTGGNSPI CYLKAYHTNL YLSADAEKVR EAIDEGIAAA TIFSPSRDVV VSQSQLRVAF DGDAVLFSDE SERIVKAHGL DRFFEHEKAH ENKPLAQGPL KGFLEALGRL QKKFYSKGLR LECPIRTYLV TARSAASSGA RALKTLRSWG LETDEALFLA GAPKGPLLEK IRPHIFFDDQ MFHVAGAQEM GTVAAHVPYG VAQTPRRTAP AKQAPSAQ | |
Greater than 90% as determined by SDS-PAGE. |
84618 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |