missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cytochrome b-c1 complex subunit 9 Protein

A DNA sequence encoding the Homo sapiens (Human) Cytochrome b-c1 complex subunit 9, was expressed in the hosts and tags indicated.

1820.00 DKK - 24645.00 DKK

Specifications

Accession Number NP_001003684.1
Gene ID (Entrez) 29796
Name Cytochrome b-c1 complex subunit 9 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16053431
View Documents
enQuireBio™
QP8232-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16093431
View Documents
enQuireBio™
QP8232-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16043431
View Documents
enQuireBio™
QP8232-ec-100ug
100 ÎĽg
5555.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16073431
View Documents
enQuireBio™
QP8232-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16083431
View Documents
enQuireBio™
QP8232-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16063431
View Documents
enQuireBio™
QP8232-ec-1mg
1 mg
24645.00 DKK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Spécification

Spécification

NP_001003684.1
Cytochrome b-c1 complex subunit 9 Protein
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
29796
Research Use Only
UQCR10
Recombinant Protein
E. coli
AAATLTSKLY SLLFRRTSTF ALTIIVGVMF FERAFDQGAD AIYDHINEGK LWKHIKHKYE NK
Greater than 80% as determined by SDS-PAGE.
Vidéos
FDS
Documents

Documents

Product Certifications
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit
enQuireBio™ Recombinant Human Cytochrome b-c1 complex subunit 9 Protein

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis